Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGIP1 SIRNA Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
UniGene ID:
Hs.132121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s38728
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308203.1
NP_001295132.1
16, 17
1369
NM_032291.3
NP_115667.2
19, 20
2146
XM_005271264.3
XP_005271321.1
19, 20
1980
XM_005271268.3
XP_005271325.1
17, 18
1395
XM_005271270.4
XP_005271327.1
16, 17
1702
XM_006710961.2
XP_006711024.1
19, 20
1986
XM_006710966.2
XP_006711029.1
18, 19
1920
XM_006710967.2
XP_006711030.1
18, 19
1908
XM_006710969.2
XP_006711032.1
18, 19
1896
XM_006710971.2
XP_006711034.1
18, 19
1485
XM_006710972.2
XP_006711035.1
18, 19
1479
XM_006710973.2
XP_006711036.1
16, 17
1461
XM_006710974.2
XP_006711037.1
15, 16
1377
XM_011542291.1
XP_011540593.1
18, 19
1642
XM_011542292.1
XP_011540594.1
17, 18
1582
XM_011542293.1
XP_011540595.1
14, 15
1311
XM_017002505.1
XP_016857994.1
20, 21
2034
XM_017002506.1
XP_016857995.1
21, 22
2157
XM_017002507.1
XP_016857996.1
20, 21
2209
XM_017002508.1
XP_016857997.1
21, 22
2155
XM_017002509.1
XP_016857998.1
18, 19
2010
XM_017002510.1
XP_016857999.1
19, 20
2133
XM_017002511.1
XP_016858000.1
20, 21
2091
XM_017002512.1
XP_016858001.1
20, 21
1964
XM_017002513.1
XP_016858002.1
19, 20
2143
XM_017002514.1
XP_016858003.1
19, 20
1962
XM_017002515.1
XP_016858004.1
17, 18
1938
XM_017002516.1
XP_016858005.1
18, 19
2061
XM_017002517.1
XP_016858006.1
19, 20
2030
XM_017002518.1
XP_016858007.1
19, 20
2018
XM_017002519.1
XP_016858008.1
19, 20
2043
XM_017002520.1
XP_016858009.1
17, 18
1884
XM_017002521.1
XP_016858010.1
17, 18
1836
XM_017002522.1
XP_016858011.1
18, 19
1958
XM_017002523.1
XP_016858012.1
18, 19
1982
XM_017002524.1
XP_016858013.1
18, 19
1625
XM_017002525.1
XP_016858014.1
17, 18
1564
XM_017002526.1
XP_016858015.1
16, 17
1443
XM_017002527.1
XP_016858016.1
17, 18
1467
XM_017002528.1
XP_016858017.1
17, 18
1553
XM_017002529.1
XP_016858018.1
17, 18
1419
XM_017002530.1
XP_016858019.1
16, 17
1395
XM_017002531.1
XP_016858020.1
15, 16
1395
XM_017002532.1
XP_016858021.1
16, 17
1389
XM_017002533.1
XP_016858022.1
16, 17
1497
XM_017002534.1
XP_016858023.1
15, 16
1371
XM_017002535.1
XP_016858024.1
16, 17
1485
XM_017002536.1
XP_016858025.1
16, 17
1335
XM_017002537.1
XP_016858026.1
15, 16
1424
GenBank mRNA
AB210039.1
-
19, 20
1928
AK090697.1
-
10, 11
926
AK125044.1
-
2, 3
913
AK298970.1
-
16, 17
1332
AK299941.1
-
17, 18
1406
AL136561.1
-
19, 20
1820
BC040516.1
-
21, 22
1974
5275011
BX640813.1
-
16, 17
1346
CR749541.1
-
16, 17
1557
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0