Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGIP1 SIRNA Assays ›
Gene Symbol:
SGIP1
Gene Name:
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 66533272 - 66751139 on Build GRCh38
UniGene ID:
Hs.132121
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s38729
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308203.1
NP_001295132.1
19
1740
NM_032291.3
NP_115667.2
22
2517
XM_005271264.3
XP_005271321.1
22
2351
XM_005271268.3
XP_005271325.1
20
1766
XM_005271270.4
XP_005271327.1
19
2073
XM_006710961.2
XP_006711024.1
22
2357
XM_006710966.2
XP_006711029.1
21
2291
XM_006710967.2
XP_006711030.1
21
2279
XM_006710969.2
XP_006711032.1
21
2267
XM_006710971.2
XP_006711034.1
21
1856
XM_006710972.2
XP_006711035.1
21
1850
XM_006710973.2
XP_006711036.1
19
1832
XM_006710974.2
XP_006711037.1
18
1748
XM_011542291.1
XP_011540593.1
21
2013
XM_011542292.1
XP_011540594.1
20
1953
XM_011542293.1
XP_011540595.1
17
1682
XM_017002505.1
XP_016857994.1
23
2405
XM_017002506.1
XP_016857995.1
24
2528
XM_017002507.1
XP_016857996.1
23
2580
XM_017002508.1
XP_016857997.1
24
2526
XM_017002509.1
XP_016857998.1
21
2381
XM_017002510.1
XP_016857999.1
22
2504
XM_017002511.1
XP_016858000.1
23
2462
XM_017002512.1
XP_016858001.1
23
2335
XM_017002513.1
XP_016858002.1
22
2514
XM_017002514.1
XP_016858003.1
22
2333
XM_017002515.1
XP_016858004.1
20
2309
XM_017002516.1
XP_016858005.1
21
2432
XM_017002517.1
XP_016858006.1
22
2401
XM_017002518.1
XP_016858007.1
22
2389
XM_017002519.1
XP_016858008.1
22
2414
XM_017002520.1
XP_016858009.1
20
2255
XM_017002521.1
XP_016858010.1
20
2207
XM_017002522.1
XP_016858011.1
21
2329
XM_017002523.1
XP_016858012.1
21
2353
XM_017002524.1
XP_016858013.1
21
1996
XM_017002525.1
XP_016858014.1
20
1935
XM_017002526.1
XP_016858015.1
19
1814
XM_017002527.1
XP_016858016.1
20
1838
XM_017002528.1
XP_016858017.1
20
1924
XM_017002529.1
XP_016858018.1
20
1790
XM_017002530.1
XP_016858019.1
19
1766
XM_017002531.1
XP_016858020.1
18
1766
XM_017002532.1
XP_016858021.1
19
1760
XM_017002533.1
XP_016858022.1
19
1868
XM_017002534.1
XP_016858023.1
18
1742
XM_017002535.1
XP_016858024.1
19
1856
XM_017002536.1
XP_016858025.1
19
1706
XM_017002537.1
XP_016858026.1
18
1795
GenBank mRNA
AB210039.1
-
22
2299
AK090697.1
-
13
1297
AK125044.1
-
5
1284
AK298970.1
-
19
1703
AK299941.1
-
20
1777
AL136561.1
-
22
2191
BC040516.1
-
24
2345
5275011
BX640813.1
-
19
1717
CR749541.1
-
19
1928
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
positive regulation of receptor-mediated endocytosis
membrane tubulation
protein binding
phospholipid binding
microtubule binding
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-s6kwc:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0