Search
Search

MEGF10| Gene Symbol: MEGF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256545.1 | NP_001243474.1 | 20 | 3018 | ||
| NM_032446.2 | NP_115822.1 | 21 | 3069 | |||
| XM_011543694.1 | XP_011541996.1 | 20 | 2781 | |||
| XM_017009987.1 | XP_016865476.1 | 22 | 3314 | |||
| XM_017009988.1 | XP_016865477.1 | 13 | 2087 | |||
| GenBank mRNA | AB058676.1 | - | 21 | 2910 | ||
| BC152478.1 | - | 21 | 2910 | 8862616 | ||
| CR749437.1 | - | 20 | 2952 | |||