Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032561.4 | NP_115950.3 | 3 | 332 | ||
XM_005261781.1 | XP_005261838.1 | 3 | 296 | |||
XM_005261782.3 | XP_005261839.1 | 3 | 176 | |||
GenBank mRNA | AF324466.1 | - | 2 | 132 | ||
AK057349.1 | - | 3 | 382 | |||
BC031998.1 | - | 3 | 214 | 4755328 | ||
BI463610.1 | - | 3 | 337 | |||
CR456440.1 | - | 3 | 212 |