Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032588.3 | NP_115977.2 | 2 | 390 | ||
XM_017002559.1 | XP_016858048.1 | 2 | 390 | |||
GenBank mRNA | AF353673.1 | - | 2 | 369 | ||
AJ276484.1 | - | 2 | 373 | |||
AK056942.1 | - | 2 | 386 | |||
AK297820.1 | - | 2 | 386 | |||
BC080529.1 | - | 2 | 368 | 4862321 | ||
BT007373.1 | - | 2 | 250 | |||
DA900348.1 | - | 2 | 390 |