Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032595.4 | NP_115984.3 | 3, 4 | 1634 | ||
XM_011525381.2 | XP_011523683.1 | 3, 4 | 2144 | |||
GenBank mRNA | AJ401189.1 | - | 3, 4 | 1624 | ||
AK054907.1 | - | 1, 2 | 107 | |||
AL713642.1 | - | 3, 4 | 441 | |||
BC016162.1 | - | 1, 2 | 121 |