Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PSRC1 SIRNA Assays ›
Gene Symbol:
PSRC1
Gene Name:
proline and serine rich coiled-coil 1
Gene Aliases:
DDA3, FP3214
Chromosome Location:
Chr.1: 109279554 - 109283182 on Build GRCh38
UniGene ID:
Hs.405925
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s39383
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PSRC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005290.3
NP_001005290.1
6
875
NM_001032291.2
NP_001027462.1
6
981
NM_032636.7
NP_116025.1
6
976
XM_005271283.2
XP_005271340.1
6
975
XM_011542306.2
XP_011540608.1
5
1235
XM_017002560.1
XP_016858049.1
5
1076
XM_017002561.1
XP_016858050.1
5
1070
XM_017002562.1
XP_016858051.1
5
1100
XM_017002563.1
XP_016858052.1
5
1143
XM_017002564.1
XP_016858053.1
5
1074
XM_017002565.1
XP_016858054.1
5
1066
XM_017002566.1
XP_016858055.1
5
1091
XM_017002567.1
XP_016858056.1
5
1086
XM_017002568.1
XP_016858057.1
5
1283
XM_017002569.1
XP_016858058.1
6
988
XM_017002570.1
XP_016858059.1
6
992
XM_017002571.1
XP_016858060.1
6
938
XM_017002572.1
XP_016858061.1
6
932
XM_017002573.1
XP_016858062.1
6
962
XM_017002574.1
XP_016858063.1
6
935
XM_017002575.1
XP_016858064.1
6
945
XM_017002576.1
XP_016858065.1
6
884
XM_017002577.1
XP_016858066.1
5
1182
GenBank mRNA
AF223000.1
-
6
955
AF447874.1
-
3
1119
AK126567.1
-
6
811
AK289593.1
-
6
918
BC056909.1
-
5
1084
6163125
BX649139.1
-
6
822
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule bundle formation
mitotic metaphase plate congression
negative regulation of cell growth
positive regulation of microtubule polymerization
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of transcription, DNA-templated
cell division
regulation of mitotic spindle organization
protein binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2gfbh:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0