Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HAVCR2 SIRNA Assays ›
          Gene Symbol:
          HAVCR2
          Gene Name:
          hepatitis A virus cellular receptor 2
          Gene Aliases:
          CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3
          Chromosome Location:
          Chr.5: 157085832 - 157109237 on Build GRCh38
          UniGene ID:
          Hs.710500
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s39535

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HAVCR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032782.4 NP_116171.3 7 1045
          GenBank mRNA AF251707.1 - 7 917
          AF450242.1 - 7 848
          AF450243.1 - 7 848
          AK027334.1 - 7 834
          AK130144.1 - 5 342
          AK314406.1 - 7 852
          AY069944.1 - 7 791
          BC063431.1 - 7 935 5229539
          JX049979.1 - 7 791

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          macrophage activation involved in immune response
          natural killer cell tolerance induction
          regulation of tolerance induction dependent upon immune response
          negative regulation of T-helper 1 type immune response
          negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
          inflammatory response
          negative regulation of gene expression
          negative regulation of myeloid dendritic cell activation
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of interferon-alpha production
          negative regulation of interferon-gamma production
          negative regulation of interleukin-2 production
          negative regulation of interleukin-3 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of chemokine production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 production
          positive regulation of interleukin-4 production
          negative regulation of natural killer cell activation
          toll-like receptor 3 signaling pathway
          toll-like receptor 7 signaling pathway
          toll-like receptor 9 signaling pathway
          positive regulation of T cell proliferation
          negative regulation of T cell proliferation
          positive regulation of macrophage activation
          innate immune response
          defense response to Gram-positive bacterium
          maternal process involved in female pregnancy
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          negative regulation of granulocyte colony-stimulating factor production
          negative regulation of defense response to bacterium
          positive regulation of defense response to bacterium
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of tumor necrosis factor secretion
          negative regulation of immunological synapse formation
          negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0