Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282736.1 | NP_001269665.1 | 5 | 1259 | ||
NM_032812.8 | NP_116201.7 | 6 | 1406 | |||
XM_011519750.2 | XP_011518052.1 | 6 | 1634 | |||
GenBank mRNA | AF378757.1 | - | 6 | 778 | ||
AK027529.1 | - | 1 | 119 | |||
AK304591.1 | - | 7 | 1290 | |||
AY358486.1 | - | 6 | 1406 | |||
BC012885.1 | - | 5 | 865 | 3865073 |