Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032826.4 | NP_116215.1 | 9 | 1063 | ||
XM_011516645.2 | XP_011514947.1 | 9 | 831 | |||
GenBank mRNA | AB052892.1 | - | 9 | 1039 | ||
AJ315497.1 | - | 9 | 895 | |||
AJ971941.1 | - | 9 | 739 | |||
AK027603.1 | - | 9 | 903 | |||
AK027726.1 | - | 8, 9 | 621 | |||
AK075368.1 | - | 9 | 868 | |||
AK222874.1 | - | 9 | 912 | |||
AK307621.1 | - | 8 | 841 | |||
BC008413.1 | - | 9 | 884 | 4333393 |