Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297701.1 | NP_001284630.1 | 6 | 427 | ||
NM_032878.3 | NP_116267.3 | 5 | 348 | |||
NM_198867.1 | NP_942567.1 | 5 | 348 | |||
XM_005259357.4 | XP_005259414.1 | 5 | 395 | |||
GenBank mRNA | AB273714.1 | - | 6 | 427 | ||
AK304805.1 | - | 5 | 322 | |||
AK310130.1 | - | 5 | 322 | |||
AK310704.1 | - | 5 | 377 | |||
AK314150.1 | - | 5 | 327 | |||
AK315894.1 | - | 5 | 316 | |||
BC007601.1 | - | 5 | 349 | |||
BC105801.1 | - | 5 | 320 | |||
BM713594.1 | - | 6 | 423 |