Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR22HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028502.1 | 2 | 398 | |||
NR_028503.1 | 2 | 398 | ||||
NR_028504.1 | 3 | 481 | ||||
NR_028505.1 | 2 | 398 | ||||
GenBank mRNA | BC119720.1 | - | 3 | 389 | 40118087 | |
BC119721.1 | - | 3 | 389 | 40118088 | ||
BM755383.1 | - | 2 | 337 | |||
BX509423.1 | - | 2 | 335 | |||
BX640650.1 | - | 3 | 419 | |||
BX648321.1 | - | 2 | 335 |