Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM86B1 SIRNA Assays ›
          Gene Symbol:
          FAM86B1 FAM86B2 FAM86DP FAM86FP FAM86HP
          Gene Name:
          family with sequence similarity 86 member B1
          family with sequence similarity 86 member B2
          family with sequence similarity 86, member D, pseudogene
          family with sequence similarity 86, member A pseudogene
          family with sequence similarity 86, member A pseudogene
          Gene Aliases:
          -
          -
          FAM86D
          -
          -
          Chromosome Location:
          Chr.8: 12182104 - 12194118 on Build GRCh38
          Chromosome Location:
          Chr.8: 12424419 - 12436418 on Build GRCh38
          Chromosome Location:
          Chr.3: 75421552 - 75435115 on Build GRCh38
          Chromosome Location:
          Chr.12: 8231049 - 8242946 on Build GRCh38
          Chromosome Location:
          Chr.3: 130097782 - 130111433 on Build GRCh38
          UniGene ID:
          Hs.675799
          Hs.458413
          Hs.591652
          Hs.661749
          Hs.123633
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s39824

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM86B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083537.1 NP_001077006.1 7 1679
          NR_003494.1 6 1413
          XM_006716256.3 XP_006716319.1 8 1774
          XM_006716257.3 XP_006716320.1 7 1844
          XM_006716260.3 XP_006716323.1 6 1588
          XM_006716262.1 XP_006716325.1 6 1431
          XM_011543842.2 XP_011542144.1 6 1853
          XM_011543843.2 XP_011542145.1 7 1792
          XM_017013916.1 XP_016869405.1 7 1820
          XM_017013917.1 XP_016869406.1 6 1741
          XM_017013918.1 XP_016869407.1 6 1713
          XM_017013919.1 XP_016869408.1 5 1302
          GenBank mRNA AK098138.1 - 7 1276
          BC062615.1 - 7 1353
          BC085016.1 - 8 1382
          BC093808.1 - 1 228
          BC093810.1 - 1 228
          BC098427.1 - 7 1359
          Gene Symbol: FAM86B2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005273603.4 XP_005273660.1 7 1709
          XM_005273604.4 XP_005273661.1 6 1628
          XM_005273605.4 XP_005273662.1 6 1529
          XM_005273607.2 XP_005273664.1 6 1567
          XM_006716387.3 XP_006716450.1 7 1730
          XM_011544618.2 XP_011542920.1 8 1781
          XM_011544619.2 XP_011542921.1 7 1748
          XM_011544622.2 XP_011542924.1 7 1650
          XM_017013740.1 XP_016869229.1 6 1711
          XM_017013741.1 XP_016869230.1 5 1648
          XM_017013742.1 XP_016869231.1 5 1630
          Gene Symbol: FAM86DP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024241.1 8 1771
          GenBank mRNA BC043342.2 - 7 1495
          Gene Symbol: FAM86FP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024254.1 5 1276
          Gene Symbol: FAM86HP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024252.1 5 1387
          GenBank mRNA AK124448.1 - 3 1155

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          methylation
          methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0