Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDC SIRNA Assays ›
          Gene Symbol:
          DDC
          Gene Name:
          dopa decarboxylase
          Gene Aliases:
          AADC
          Chromosome Location:
          Chr.7: 50458436 - 50565460 on Build GRCh38
          UniGene ID:
          Hs.359698
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s3987

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000790.3 NP_000781.1 9 1005
          NM_001082971.1 NP_001076440.1 9 1120
          NM_001242886.1 NP_001229815.1 8 891
          NM_001242887.1 NP_001229816.1 8 861
          NM_001242888.1 NP_001229817.1 7 771
          NM_001242889.1 NP_001229818.1 7 726
          NM_001242890.1 NP_001229819.1 9 1005
          XM_005271745.4 XP_005271802.1 8 1009
          XM_011515161.2 XP_011513463.2 7 877
          GenBank mRNA AJ310724.1 - 9 982
          AK298321.1 - 8 836
          AK298392.1 - 8 881
          AU310260.1 - 7 495
          BC000485.1 - 9 989 2819761
          BC008366.1 - 9 994 2819761
          BT007009.1 - 8 919
          CD013934.1 - 7 754
          CD013935.1 - 7 709
          CD013936.1 - 8 874
          CD013937.1 - 8 844
          CD013938.1 - 7 779
          CR456724.1 - 8 919
          KU178021.1 - 8 919
          M76180.1 - 9 988
          M88700.1 - 9 1008

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          decarboxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular amino acid metabolic process
          circadian rhythm
          multicellular organism aging
          aminergic neurotransmitter loading into synaptic vesicle
          isoquinoline alkaloid metabolic process
          cellular response to drug
          dopamine biosynthetic process
          catecholamine biosynthetic process
          serotonin biosynthetic process
          indolalkylamine biosynthetic process
          response to pyrethroid
          phytoalexin metabolic process
          cellular response to alkaloid
          cellular response to growth factor stimulus
          aromatic-L-amino-acid decarboxylase activity
          protein binding
          amino acid binding
          enzyme binding
          protein domain specific binding
          pyridoxal phosphate binding
          L-dopa decarboxylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0