Search
Search

SLC45A3| Gene Symbol: SLC45A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033102.2 | NP_149093.1 | 5 | 2861 | ||
| XM_005245556.3 | XP_005245613.1 | 5 | 3027 | |||
| XM_005245557.4 | XP_005245614.1 | 5 | 2953 | |||
| XM_017002638.1 | XP_016858127.1 | 5 | 3400 | |||
| GenBank mRNA | AL832933.1 | - | 3 | 1925 | ||
| AY033593.1 | - | 5 | 2803 | |||
| BC050416.2 | - | 5 | 2769 | 6198823 | ||