Search
Search

DIXDC1| Gene Symbol: DIXDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037954.3 | NP_001033043.1 | 8, 9 | 1302 | ||
| NM_033425.4 | NP_219493.1 | 4, 5 | 835 | |||
| XM_017018466.1 | XP_016873955.1 | 9, 10 | 1262 | |||
| XM_017018467.1 | XP_016873956.1 | 9, 10 | 1130 | |||
| XM_017018468.1 | XP_016873957.1 | 4, 5 | 446 | |||
| XM_017018469.1 | XP_016873958.1 | 5, 6 | 471 | |||
| GenBank mRNA | AB051522.1 | - | 4, 5 | 742 | ||
| AK055899.1 | - | 2, 3 | 358 | |||
| AK289807.1 | - | 4, 5 | 811 | |||
| BC033034.2 | - | 4, 5 | 794 | 4820652 | ||
| BC041626.1 | - | 1, 2 | 29 | |||
| DQ642016.1 | - | 8, 9 | 1302 | |||