Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193416.2 | NP_001180345.1 | 5 | 471 | ||
NM_001193417.2 | NP_001180346.1 | 4 | 423 | |||
NM_001356.4 | NP_001347.3 | 5 | 1325 | |||
NR_126093.1 | 5 | 1325 | ||||
XM_011543892.2 | XP_011542194.1 | 5 | 502 | |||
XM_017029313.1 | XP_016884802.1 | 5 | 2670 | |||
XM_017029314.1 | XP_016884803.1 | 5 | 2670 | |||
GenBank mRNA | AB208983.1 | - | 5 | 423 | ||
AB451220.1 | - | 5 | 380 | |||
AB451343.1 | - | 5 | 380 | |||
AF000982.1 | - | 5 | 1236 | |||
AF000983.1 | - | 5 | 1236 | |||
AF061337.1 | - | 5 | 455 | |||
AK291153.1 | - | 5 | 471 | |||
AK297159.1 | - | 5 | 471 | |||
AK304661.1 | - | 5 | 514 | |||
AK304689.1 | - | 4 | 423 | |||
AK310891.1 | - | 4 | 423 | |||
BC011819.2 | - | 5 | 450 | 3617040 | ||
BM833870.1 | - | 5 | 471 | |||
U50553.1 | - | 5 | 689 |