Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052854.3 | NP_443086.1 | 7, 8 | 1445 | ||
XM_006718380.3 | XP_006718443.1 | 7, 8 | 1422 | |||
GenBank mRNA | AB063321.1 | - | 7, 8 | 1066 | ||
AK075538.1 | - | 2, 3 | 216 | |||
AK314072.1 | - | 7, 8 | 1423 | |||
BC014097.1 | - | 7, 8 | 1088 | 4546950 | ||
BC015781.1 | - | 7, 8 | 1074 | 4856946 |