Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220494.2 | NP_001207423.1 | 8 | 1175 | ||
NM_001220496.2 | NP_001207425.1 | 7 | 1003 | |||
NM_080661.4 | NP_542392.2 | 7 | 906 | |||
XM_011545354.2 | XP_011543656.1 | 7 | 1022 | |||
XM_011545355.2 | XP_011543657.1 | 7 | 1008 | |||
XM_011545356.2 | XP_011543658.1 | 8 | 1067 | |||
XM_011545357.2 | XP_011543659.1 | 8 | 1053 | |||
XM_011545358.2 | XP_011543660.1 | 8 | 1591 | |||
XM_011545359.2 | XP_011543661.1 | 7 | 1547 | |||
GenBank mRNA | AB207212.1 | - | 5 | 763 | ||
AK055818.1 | - | 6 | 1160 | |||
AK091965.1 | - | 7 | 906 | |||
AK130017.1 | - | 4 | 1760 | |||
AK300479.1 | - | 8 | 1071 | |||
BC008353.1 | - | 8 | 1103 | 3537224 | ||
DQ084381.1 | - | 7 | 1003 |