Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145235.3 | NP_660278.3 | 2, 3 | 289 | ||
XM_006718058.3 | XP_006718121.1 | 3, 4 | 270 | |||
XM_006718064.3 | XP_006718127.1 | 2, 3 | 264 | |||
XM_011540348.2 | XP_011538650.1 | 3, 4 | 325 | |||
XM_011540349.2 | XP_011538651.1 | 3, 4 | 329 | |||
XM_011540350.2 | XP_011538652.1 | 2, 3 | 288 | |||
XM_011540351.2 | XP_011538653.1 | 2, 3 | 264 | |||
XM_011540356.2 | XP_011538658.1 | 2, 3 | 254 | |||
XM_011540357.2 | XP_011538659.1 | 3, 4 | 272 | |||
XM_011540358.2 | XP_011538660.1 | 2, 3 | 269 | |||
XM_011540363.2 | XP_011538665.1 | 4, 5 | 488 | |||
XM_017016916.1 | XP_016872405.1 | 3, 4 | 271 | |||
XM_017016917.1 | XP_016872406.1 | 3, 4 | 406 | |||
XM_017016918.1 | XP_016872407.1 | 2, 3 | 213 | |||
XM_017016919.1 | XP_016872408.1 | 2, 3 | 365 | |||
XM_017016920.1 | XP_016872409.1 | 4, 5 | 488 | |||
XM_017016921.1 | XP_016872410.1 | 3, 4 | 406 | |||
XM_017016923.1 | XP_016872412.1 | 2, 3 | 288 | |||
GenBank mRNA | AK304407.1 | - | 3, 4 | 340 | ||
AY251163.1 | - | 1, 2 | 189 | |||
AY358154.1 | - | 2, 3 | 267 | |||
BC024189.2 | - | 2, 3 | 289 | 4826296 | ||
BQ225468.1 | - | 2, 3 | 233 |