Hamburger Menu Button
Thermo Fisher Scientific Logo
登录
没有账号?
建立账号
注册
产品
抗体
Custom DNA Oligos
GeneArt Gene Synthesis Instant Designer
TaqMan Assay and Array Search
PCR 设备和耗材
色谱柱和纯化柱
实验室离心机
分子生物学试剂和试剂盒
移液器和移液器吸头
查看所有产品分类
应用和技术
色谱
细胞转染和培养
实时荧光定量PCR
质谱
实验室塑料耗材及用品
电子显微镜
蛋白质生物学
PCR
RNAi
环境
查看所有应用和技术
服务
定制服务
企业级实验室信息化服务
企业服务
金融&租赁服务
仪器服务
OEM和商业供应
培训服务
药物开发生产和临床实验服务
查看所有帮助和支持
帮助和支持
学习中心
联系我们
更改国家或地区
查看所有帮助和支持
联系我们
货号直购
订单查询
查找文件&证书
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
搜索全部或下列指定分类
Search
Search button
Search button
Close
联系我们
货号直购
登录
登录
没有账号?
建立账号
注册
我的账号
订单
定制产品及项目
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPH7 SIRNA Assays ›
Gene Symbol:
DPH7
Gene Name:
diphthamide biosynthesis 7
Gene Aliases:
C9orf112, RRT2, WDR85
Chromosome Location:
Chr.9: 137554899 - 137578935 on Build GRCh38
UniGene ID:
Hs.292570
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41004
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
添加到购物车
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138778.3
NP_620133.1
9
1228
XM_005266123.2
XP_005266180.1
10
1686
XM_005266126.3
XP_005266183.1
8
1459
XM_005266128.2
XP_005266185.1
10
1312
XM_005266129.2
XP_005266186.1
10
1315
XM_005266130.2
XP_005266187.1
8
1083
XM_011519188.1
XP_011517490.1
8
1151
XM_011519189.1
XP_011517491.1
9
1329
XM_011519191.2
XP_011517493.1
9
2384
XM_011519193.2
XP_011517495.1
9
2433
XM_011519199.2
XP_011517501.1
9
1174
XM_011519200.1
XP_011517502.1
9
1177
XM_017015289.1
XP_016870778.1
8
2318
XM_017015290.1
XP_016870779.1
8
2367
XM_017015291.1
XP_016870780.1
10
1521
XM_017015292.1
XP_016870781.1
9
1591
XM_017015293.1
XP_016870782.1
9
1548
XM_017015294.1
XP_016870783.1
11
1501
XM_017015295.1
XP_016870784.1
9
1588
XM_017015296.1
XP_016870785.1
9
1427
XM_017015297.1
XP_016870786.1
11
1498
XM_017015298.1
XP_016870787.1
10
1781
XM_017015299.1
XP_016870788.1
10
1038
XM_017015300.1
XP_016870789.1
10
1778
XM_017015301.1
XP_016870790.1
8
1451
XM_017015302.1
XP_016870791.1
8
2505
XM_017015303.1
XP_016870792.1
9
1686
XM_017015304.1
XP_016870793.1
8
1151
XM_017015305.1
XP_016870794.1
8
2482
XM_017015306.1
XP_016870795.1
9
1620
XM_017015307.1
XP_016870796.1
9
1617
XM_017015308.1
XP_016870797.1
7
2344
XM_017015309.1
XP_016870798.1
9
1344
GenBank mRNA
AJ420445.1
-
1
587
AK075115.1
-
9
1229
AK094389.1
-
1
1229
BC003123.1
-
9
1167
3502107
BC017335.2
-
9
1187
4642600
BC040173.1
-
2
690
4791980
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
diphthine methylesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-fwnls:80/10.239.167.40:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0