Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145335.1 | NP_001138807.1 | 12 | 1324 | ||
NM_001284382.1 | NP_001271311.1 | 11 | 1338 | |||
NM_173469.3 | NP_775740.1 | 12 | 1443 | |||
XM_005254788.1 | XP_005254845.1 | 9 | 978 | |||
XM_017022728.1 | XP_016878217.1 | 8 | 901 | |||
GenBank mRNA | AK000617.1 | - | 11 | 1317 | ||
AK294199.1 | - | 12 | 984 | |||
AK296234.1 | - | 12 | 1324 | |||
AL832429.1 | - | 12 | 1268 | |||
BC017708.1 | - | 12 | 1270 | 3907760 | ||
BC034342.1 | - | 4 | 236 |