Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TJAP1 SIRNA Assays ›
Gene Symbol:
TJAP1
Gene Name:
tight junction associated protein 1
Gene Aliases:
PILT, TJP4
Chromosome Location:
Chr.6: 43477523 - 43506557 on Build GRCh38
UniGene ID:
Hs.520145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41180
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TJAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146016.1
NP_001139488.1
9
779
NM_001146017.1
NP_001139489.1
8
744
NM_001146018.1
NP_001139490.1
9
806
NM_001146019.1
NP_001139491.1
8
749
NM_001146020.1
NP_001139492.1
7
600
NM_080604.2
NP_542171.2
8
801
NR_027394.1
8
666
XM_006715249.1
XP_006715312.1
9
757
XM_006715250.3
XP_006715313.1
10
776
XM_006715251.2
XP_006715314.1
9
784
XM_006715252.2
XP_006715315.1
10
844
XM_006715254.3
XP_006715317.1
9
682
XM_006715256.1
XP_006715319.1
9
679
XM_006715257.2
XP_006715320.1
9
733
XM_006715259.3
XP_006715322.1
8
583
XM_006715260.1
XP_006715323.1
9
700
XM_006715261.1
XP_006715324.1
8
604
XM_006715262.2
XP_006715325.1
9
929
XM_006715263.2
XP_006715326.1
8
639
XM_006715265.1
XP_006715328.1
8
649
XM_006715266.1
XP_006715329.1
7
557
XM_006715267.2
XP_006715330.1
9
768
XM_006715268.2
XP_006715331.1
9
765
XM_006715269.2
XP_006715332.1
9
820
XM_006715270.1
XP_006715333.1
8
738
XM_011514995.1
XP_011513297.1
10
819
XM_011514996.1
XP_011513298.1
9
739
XM_017011488.1
XP_016866977.1
8
657
XM_017011489.1
XP_016866978.1
9
954
XM_017011490.1
XP_016866979.1
8
727
XM_017011491.1
XP_016866980.1
9
763
XM_017011492.1
XP_016866981.1
10
892
XM_017011493.1
XP_016866982.1
10
890
GenBank mRNA
AK024269.1
-
8
751
AK074241.1
-
3
195
AK094907.1
-
1
607
AK123398.1
-
5
3309
AK127338.1
-
7
1862
AK225787.1
-
8
702
AK309119.1
-
8
619
AL117398.1
-
3
179
BC046239.1
-
9
699
4906152
BC064401.1
-
8
680
5539930
CR749480.1
-
6
2170
DA051961.1
-
7
553
DA297698.1
-
8
551
Back To Top
More Information
Panther Classification:
Molecular Function -
tight junction
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0