Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LARP4 SIRNA Assays ›
          Gene Symbol:
          LARP4
          Gene Name:
          La ribonucleoprotein domain family member 4
          Gene Aliases:
          PP13296
          Chromosome Location:
          Chr.12: 50400809 - 50480005 on Build GRCh38
          UniGene ID:
          Hs.26613
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s41456

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LARP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001170803.1 NP_001164274.1 11 1205
          NM_001170808.1 NP_001164279.1 12 1418
          NM_052879.4 NP_443111.4 13 1631
          NM_199188.2 NP_954658.2 13 1628
          NM_199190.2 NP_954660.1 12 1418
          XM_005268613.2 XP_005268670.1 14 1680
          XM_011537828.2 XP_011536130.1 14 1735
          XM_011537829.2 XP_011536131.1 14 1716
          XM_011537830.2 XP_011536132.1 13 1563
          XM_011537831.2 XP_011536133.1 13 1627
          XM_011537833.2 XP_011536135.1 13 1522
          XM_011537834.2 XP_011536136.1 13 1523
          XM_011537835.2 XP_011536137.1 13 1503
          XM_011537836.2 XP_011536138.1 13 1504
          XM_011537838.2 XP_011536140.1 13 1475
          XM_011537839.2 XP_011536141.1 14 1619
          XM_011537840.2 XP_011536142.1 14 1672
          XM_011537841.2 XP_011536143.1 13 1525
          XM_011537843.2 XP_011536145.1 12 1291
          XM_011537844.2 XP_011536146.1 14 1736
          XM_017018750.1 XP_016874239.1 12 1344
          XM_017018751.1 XP_016874240.1 14 1496
          XM_017018752.1 XP_016874241.1 13 1479
          XM_017018753.1 XP_016874242.1 13 1512
          XM_017018754.1 XP_016874243.1 14 1662
          XM_017018755.1 XP_016874244.1 12 1329
          XM_017018756.1 XP_016874245.1 13 1381
          XM_017018757.1 XP_016874246.1 14 1551
          XM_017018758.1 XP_016874247.1 13 1487
          XM_017018759.1 XP_016874248.1 12 1312
          XM_017018760.1 XP_016874249.1 13 1631
          XM_017018761.1 XP_016874250.1 12 1411
          XM_017018762.1 XP_016874251.1 12 1411
          GenBank mRNA AF370416.1 - 13 1509
          AK055521.1 - 11 1179
          AK125113.1 - 12 1421
          AK291746.1 - 13 1500
          AL832198.1 - 12 1359
          AL833272.1 - 13 1541
          BC022377.1 - 11 1105
          BX647457.1 - 12 1242
          CR936626.1 - 13 1549

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          regulation of cell morphogenesis
          nucleotide binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0