Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP4 SIRNA Assays ›
Gene Symbol:
LARP4
Gene Name:
La ribonucleoprotein domain family member 4
Gene Aliases:
PP13296
Chromosome Location:
Chr.12: 50400809 - 50480005 on Build GRCh38
UniGene ID:
Hs.26613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41457
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170803.1
NP_001164274.1
13
1500
NM_001170808.1
NP_001164279.1
14
1713
NM_052879.4
NP_443111.4
15
1926
NM_199188.2
NP_954658.2
15
1923
NM_199190.2
NP_954660.1
14
1713
XM_005268613.2
XP_005268670.1
16
1975
XM_011537828.2
XP_011536130.1
16
2030
XM_011537829.2
XP_011536131.1
16
2011
XM_011537830.2
XP_011536132.1
15
1858
XM_011537831.2
XP_011536133.1
15
1922
XM_011537833.2
XP_011536135.1
15
1817
XM_011537834.2
XP_011536136.1
15
1818
XM_011537835.2
XP_011536137.1
15
1798
XM_011537836.2
XP_011536138.1
15
1799
XM_011537838.2
XP_011536140.1
15
1770
XM_011537839.2
XP_011536141.1
16
1914
XM_011537840.2
XP_011536142.1
16
1967
XM_011537841.2
XP_011536143.1
15
1820
XM_011537843.2
XP_011536145.1
14
1586
XM_011537844.2
XP_011536146.1
15
1909
XM_017018750.1
XP_016874239.1
14
1639
XM_017018751.1
XP_016874240.1
16
1791
XM_017018752.1
XP_016874241.1
15
1774
XM_017018753.1
XP_016874242.1
15
1807
XM_017018754.1
XP_016874243.1
16
1957
XM_017018755.1
XP_016874244.1
14
1624
XM_017018756.1
XP_016874245.1
15
1676
XM_017018757.1
XP_016874246.1
16
1846
XM_017018758.1
XP_016874247.1
15
1782
XM_017018759.1
XP_016874248.1
14
1607
XM_017018760.1
XP_016874249.1
14
1804
XM_017018761.1
XP_016874250.1
13
1584
XM_017018762.1
XP_016874251.1
13
1584
GenBank mRNA
AF370416.1
-
15
1804
AK055521.1
-
13
1474
AK125113.1
-
14
1716
AK291746.1
-
15
1795
AL832198.1
-
14
1654
AL833272.1
-
15
1836
BC022377.1
-
13
1400
BC032444.2
-
2
181
BX647457.1
-
14
1537
CR936626.1
-
15
1844
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-njpwx:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0