Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DMD SIRNA Assays ›
          Gene Symbol:
          DMD
          Gene Name:
          dystrophin
          Gene Aliases:
          BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
          Chromosome Location:
          Chr.X: 31119219 - 33339609 on Build GRCh38
          UniGene ID:
          Hs.495912
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4155

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DMD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000109.3 NP_000100.2 63, 64 9607
          NM_004006.2 NP_003997.1 63, 64 9531
          NM_004009.3 NP_004000.1 63, 64 9538
          NM_004010.3 NP_004001.1 63, 64 9621
          NM_004011.3 NP_004002.2 35, 36 5309
          NM_004012.3 NP_004003.1 35, 36 5452
          NM_004013.2 NP_004004.1 20, 21 2948
          NM_004014.2 NP_004005.1 9, 10 1161
          NM_004015.2 NP_004006.1 2, 3 161
          NM_004016.2 NP_004007.1 2, 3 161
          NM_004017.2 NP_004008.1 2, 3 161
          NM_004018.2 NP_004009.1 2, 3 161
          NM_004019.2 NP_004010.1 2, 3 161
          NM_004020.3 NP_004011.2 20, 21 2948
          NM_004021.2 NP_004012.1 20, 21 2948
          NM_004022.2 NP_004013.1 20, 21 2948
          NM_004023.2 NP_004014.1 20, 21 2948
          XM_006724468.2 XP_006724531.1 63, 64 9484
          XM_006724469.3 XP_006724532.1 63, 64 9458
          XM_006724470.3 XP_006724533.1 63, 64 9484
          XM_006724473.2 XP_006724536.1 62, 63 9346
          XM_006724474.3 XP_006724537.1 63, 64 9484
          XM_006724475.2 XP_006724538.1 63, 64 9484
          XM_011545467.1 XP_011543769.1 62, 63 9361
          XM_011545468.2 XP_011543770.1 63, 64 9484
          XM_017029328.1 XP_016884817.1 63, 64 9484
          XM_017029331.1 XP_016884820.1 23, 24 4060
          GenBank mRNA AB208836.1 - 17, 18 2395
          AK299936.1 - 2, 3 203
          BC028720.1 - 2, 3 212
          BC070078.1 - 2, 3 162
          BC094758.1 - 2, 3 175
          BC127103.1 - 17, 18 2472
          BC150141.1 - 17, 18 2472
          DB233002.1 - 2, 3 161
          KU178030.1 - 2, 3 83
          KU178031.1 - 2, 3 83
          KU178032.1 - 2, 3 83
          KU178033.1 - 2, 3 83
          KU178034.1 - 2, 3 83
          M18533.1 - 63, 64 9495
          M92650.1 - 2, 3 135
          S38776.1 - 2, 3 206
          X14298.1 - 63, 64 9385

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001005246, XM_008773253, XM_008773252, X65468, X69767, and others.
          This siRNA also targets the following transcripts in Mouse: NM_007868, KJ480729, S62620, KJ480728, M68859, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          cytoskeleton organization
          muscle organ development
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
          regulation of skeletal muscle contraction
          muscle attachment
          muscle filament sliding
          negative regulation of peptidyl-serine phosphorylation
          cellular protein localization
          response to muscle stretch
          peptide biosynthetic process
          cellular protein complex assembly
          muscle cell cellular homeostasis
          muscle fiber development
          cardiac muscle contraction
          regulation of ryanodine-sensitive calcium-release channel activity
          cardiac muscle cell action potential
          regulation of voltage-gated calcium channel activity
          negative regulation of peptidyl-cysteine S-nitrosylation
          positive regulation of sodium ion transmembrane transporter activity
          dystroglycan binding
          actin binding
          structural constituent of cytoskeleton
          protein binding
          zinc ion binding
          structural constituent of muscle
          myosin binding
          vinculin binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0