Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMD SIRNA Assays ›
Gene Symbol:
DMD
Gene Name:
dystrophin
Gene Aliases:
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
Chromosome Location:
Chr.X: 31119219 - 33339609 on Build GRCh38
UniGene ID:
Hs.495912
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4156
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000109.3
NP_000100.2
67
10046
NM_004006.2
NP_003997.1
67
9970
NM_004009.3
NP_004000.1
67
9977
NM_004010.3
NP_004001.1
67
10060
NM_004011.3
NP_004002.2
39
5748
NM_004012.3
NP_004003.1
39
5891
NM_004013.2
NP_004004.1
24
3387
NM_004014.2
NP_004005.1
13
1600
NM_004015.2
NP_004006.1
6
600
NM_004016.2
NP_004007.1
6
600
NM_004017.2
NP_004008.1
6
600
NM_004018.2
NP_004009.1
6
600
NM_004019.2
NP_004010.1
6
600
NM_004020.3
NP_004011.2
24
3387
NM_004021.2
NP_004012.1
24
3387
NM_004022.2
NP_004013.1
24
3387
NM_004023.2
NP_004014.1
24
3387
XM_006724468.2
XP_006724531.1
67
9923
XM_006724469.3
XP_006724532.1
67
9897
XM_006724470.3
XP_006724533.1
67
9923
XM_006724473.2
XP_006724536.1
66
9785
XM_006724474.3
XP_006724537.1
67
9923
XM_006724475.2
XP_006724538.1
67
9923
XM_011545467.1
XP_011543769.1
66
9800
XM_011545468.2
XP_011543770.1
67
9923
XM_017029328.1
XP_016884817.1
67
9923
XM_017029331.1
XP_016884820.1
27
4499
GenBank mRNA
AB208836.1
-
21
2834
AK299936.1
-
6
642
BC028720.1
-
6
651
BC070078.1
-
6
601
BC094758.1
-
6
614
BC127103.1
-
21
2911
BC150141.1
-
21
2911
KU178030.1
-
6
522
KU178031.1
-
6
522
KU178032.1
-
6
522
KU178033.1
-
6
522
M18533.1
-
67
9934
M92650.1
-
6
574
X14298.1
-
67
9824
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cytoskeleton organization
muscle organ development
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
muscle attachment
muscle filament sliding
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
peptide biosynthetic process
cellular protein complex assembly
muscle cell cellular homeostasis
muscle fiber development
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
cardiac muscle cell action potential
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
structural constituent of cytoskeleton
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0