Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRRC37B SIRNA Assays ›
          Gene Symbol:
          LRRC37B LRRC37A2 LRRC37A4P
          Gene Name:
          leucine rich repeat containing 37B
          leucine rich repeat containing 37 member A2
          leucine rich repeat containing 37 member A4, pseudogene
          Gene Aliases:
          LRRC37
          LRRC37
          LRRC37, LRRC37A4
          Chromosome Location:
          Chr.17: 32007872 - 32053500 on Build GRCh38
          Chromosome Location:
          Chr.17: 46512657 - 46556747 on Build GRCh38
          Chromosome Location:
          Chr.17: 45505883 - 45520523 on Build GRCh38
          UniGene ID:
          Hs.514071
          Hs.646568
          Hs.596756
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s41567

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRRC37B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321350.1 NP_001308279.1 11, 12 2442
          NM_052888.2 NP_443120.2 8, 9 2248
          GenBank mRNA AJ314647.1 - 8, 9 2234
          AK299772.1 - 11, 12 2296
          AK302740.1 - 8, 9 2379
          AY386261.1 - 9, 10 2286
          BC117373.1 - 6, 7 2134 40125924
          BC143581.1 - 6, 7 2134 9052092
          JQ790561.1 - 8, 9 719
          JQ790565.1 - 8, 9 727
          JQ790567.1 - 8, 9 727
          JQ790569.1 - 8, 9 727
          JQ790582.1 - 8, 9 727
          JQ790601.1 - 8, 9 727
          JQ790602.1 - 8, 9 727
          JQ790604.1 - 8, 9 727
          JQ790615.1 - 8, 9 727
          JQ790616.1 - 8, 9 727
          JQ790618.1 - 8, 9 727
          JQ790619.1 - 8, 9 727
          JQ790620.1 - 8 727
          JQ790621.1 - 8, 9 727
          JQ790622.1 - 8 715
          JQ790672.1 - 5, 6 502
          JQ790687.1 - 8 727
          JQ790695.1 - 8, 9 727
          JQ790696.1 - 8, 9 727
          JQ790701.1 - 8, 9 727
          JQ790746.1 - 8, 9 729
          JQ790754.1 - 8 729
          Gene Symbol: LRRC37A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA JQ790576.1 - 8, 9 727
          Gene Symbol: LRRC37A4P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA JQ790698.1 - 8, 9 727

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0