Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DMD SIRNA Assays ›
          Gene Symbol:
          DMD
          Gene Name:
          dystrophin
          Gene Aliases:
          BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
          Chromosome Location:
          Chr.X: 31119219 - 33339609 on Build GRCh38
          UniGene ID:
          Hs.495912
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4157

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DMD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000109.3 NP_000100.2 66, 67 9972
          NM_004006.2 NP_003997.1 66, 67 9896
          NM_004009.3 NP_004000.1 66, 67 9903
          NM_004010.3 NP_004001.1 66, 67 9986
          NM_004011.3 NP_004002.2 38, 39 5674
          NM_004012.3 NP_004003.1 38, 39 5817
          NM_004013.2 NP_004004.1 23, 24 3313
          NM_004014.2 NP_004005.1 12, 13 1526
          NM_004015.2 NP_004006.1 5, 6 526
          NM_004016.2 NP_004007.1 5, 6 526
          NM_004017.2 NP_004008.1 5, 6 526
          NM_004018.2 NP_004009.1 5, 6 526
          NM_004019.2 NP_004010.1 5, 6 526
          NM_004020.3 NP_004011.2 23, 24 3313
          NM_004021.2 NP_004012.1 23, 24 3313
          NM_004022.2 NP_004013.1 23, 24 3313
          NM_004023.2 NP_004014.1 23, 24 3313
          XM_006724468.2 XP_006724531.1 66, 67 9849
          XM_006724469.3 XP_006724532.1 66, 67 9823
          XM_006724470.3 XP_006724533.1 66, 67 9849
          XM_006724473.2 XP_006724536.1 65, 66 9711
          XM_006724474.3 XP_006724537.1 66, 67 9849
          XM_006724475.2 XP_006724538.1 66, 67 9849
          XM_011545467.1 XP_011543769.1 65, 66 9726
          XM_011545468.2 XP_011543770.1 66, 67 9849
          XM_017029328.1 XP_016884817.1 66, 67 9849
          XM_017029331.1 XP_016884820.1 26, 27 4425
          GenBank mRNA AB208836.1 - 20, 21 2760
          AK299936.1 - 5, 6 568
          BC028720.1 - 5, 6 577
          BC070078.1 - 5, 6 527
          BC094758.1 - 5, 6 540
          BC127103.1 - 20, 21 2837
          BC150141.1 - 20, 21 2837
          DB233002.1 - 5, 6 526
          KU178030.1 - 5, 6 448
          KU178031.1 - 5, 6 448
          KU178032.1 - 5, 6 448
          KU178033.1 - 5, 6 448
          M18533.1 - 66, 67 9860
          M92650.1 - 5, 6 500
          X14298.1 - 66, 67 9750

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001005246, XM_008773253, XM_008773252, X69767.
          This siRNA also targets the following transcripts in Mouse: NM_007868, KJ480729, KJ480728, M68859, AK075809, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          cytoskeleton organization
          muscle organ development
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
          regulation of skeletal muscle contraction
          muscle attachment
          muscle filament sliding
          negative regulation of peptidyl-serine phosphorylation
          cellular protein localization
          response to muscle stretch
          peptide biosynthetic process
          cellular protein complex assembly
          muscle cell cellular homeostasis
          muscle fiber development
          cardiac muscle contraction
          regulation of ryanodine-sensitive calcium-release channel activity
          cardiac muscle cell action potential
          regulation of voltage-gated calcium channel activity
          negative regulation of peptidyl-cysteine S-nitrosylation
          positive regulation of sodium ion transmembrane transporter activity
          dystroglycan binding
          actin binding
          structural constituent of cytoskeleton
          protein binding
          zinc ion binding
          structural constituent of muscle
          myosin binding
          vinculin binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0