Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDKN1A SIRNA Assays ›
          Gene Symbol:
          CDKN1A
          Gene Name:
          cyclin dependent kinase inhibitor 1A
          Gene Aliases:
          CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
          Chromosome Location:
          Chr.6: 36676460 - 36687339 on Build GRCh38
          UniGene ID:
          Hs.370771
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s416
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDKN1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000389.4 NP_000380.1 3 1560
          NM_001220777.1 NP_001207706.1 3 1505
          NM_001220778.1 NP_001207707.1 3 1670
          NM_001291549.1 NP_001278478.1 4 1712
          NM_078467.2 NP_510867.1 4 1507
          GenBank mRNA AB209881.1 - 3 1517
          BC000275.1 - 3 1500 3355833
          BC000312.2 - 3 1654 2821049
          BC001935.1 - 3 1515 2823668
          BC013967.2 - 3 1518 2822909
          BQ941282.1 - 1 105
          L25610.1 - 3 1513
          L26165.1 - 3 1513
          U03106.1 - 3 1511
          U09579.1 - 3 1529

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00355782_m1

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase inhibitor kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          G1/S transition of mitotic cell cycle
          G2/M transition of mitotic cell cycle
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          cell cycle arrest
          Ras protein signal transduction
          negative regulation of cell proliferation
          response to toxic substance
          response to X-ray
          response to organonitrogen compound
          negative regulation of gene expression
          negative regulation of cell growth
          positive regulation of B cell proliferation
          organ regeneration
          cellular response to extracellular stimulus
          regulation of protein import into nucleus, translocation
          cellular response to amino acid starvation
          cellular response to heat
          negative regulation of phosphorylation
          response to drug
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of apoptotic process
          positive regulation of programmed cell death
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          response to arsenic-containing substance
          positive regulation of fibroblast proliferation
          protein stabilization
          response to corticosterone
          response to hyperoxia
          intestinal epithelial cell maturation
          cellular response to ionizing radiation
          cellular response to UV-B
          mitotic cell cycle arrest
          cellular senescence
          replicative senescence
          stress-induced premature senescence
          intrinsic apoptotic signaling pathway
          positive regulation of cyclin-dependent protein kinase activity
          negative regulation of G1/S transition of mitotic cell cycle
          regulation of DNA biosynthetic process
          positive regulation of reactive oxygen species metabolic process
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding
          cyclin-dependent protein kinase activating kinase activity
          cyclin binding
          ubiquitin protein ligase binding
          protein complex binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0