Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OSBPL6 SIRNA Assays ›
          Gene Symbol:
          OSBPL6
          Gene Name:
          oxysterol binding protein like 6
          Gene Aliases:
          ORP6
          Chromosome Location:
          Chr.2: 178194479 - 178399433 on Build GRCh38
          UniGene ID:
          Hs.318775
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s41685

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OSBPL6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001201480.1 NP_001188409.1 26 3352
          NM_001201481.1 NP_001188410.1 24 3184
          NM_001201482.1 NP_001188411.1 24 3169
          NM_032523.3 NP_115912.1 25 3277
          NM_145739.2 NP_665682.1 24 2865
          XM_011510539.1 XP_011508841.1 25 3255
          XM_017003265.1 XP_016858754.1 27 3424
          XM_017003266.1 XP_016858755.1 27 3424
          XM_017003267.1 XP_016858756.1 25 3119
          XM_017003268.1 XP_016858757.1 25 3240
          XM_017003269.1 XP_016858758.1 24 3011
          XM_017003270.1 XP_016858759.1 25 3338
          XM_017003271.1 XP_016858760.1 23 2936
          XM_017003272.1 XP_016858761.1 24 3147
          XM_017003273.1 XP_016858762.1 23 3087
          XM_017003274.1 XP_016858763.1 23 3072
          XM_017003275.1 XP_016858764.1 24 3166
          XM_017003276.1 XP_016858765.1 23 3072
          XM_017003277.1 XP_016858766.1 22 2979
          GenBank mRNA AB208898.1 - 26 3352
          AF323728.1 - 25 3111
          AF392448.1 - 25 3111
          AF462443.1 - 23 2979
          AK027600.1 - 18 2152
          AK093902.1 - 24 2867
          AK300389.1 - 24 3142

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107735, XM_008761902, XM_008761903, XM_008761906, XM_008761904, and others.
          This siRNA also targets the following transcripts in Mouse: NM_145525, NM_001290734, NM_001290733, AK048018, AK037703, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid transport
          protein binding
          lipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rlpgz:80/100.66.72.8:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0