Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OSBPL8 SIRNA Assays ›
          Gene Symbol:
          OSBPL8
          Gene Name:
          oxysterol binding protein like 8
          Gene Aliases:
          MST120, MSTP120, ORP8, OSBP10
          Chromosome Location:
          Chr.12: 76351797 - 76559809 on Build GRCh38
          UniGene ID:
          Hs.430849
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s41692
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OSBPL8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003712.1 NP_001003712.1 8 1131
          NM_001319652.1 NP_001306581.1 11 1256
          NM_001319653.1 NP_001306582.1 9 1168
          NM_001319655.1 NP_001306584.1 6 1062
          NM_020841.4 NP_065892.1 9 1177
          XM_005268621.4 XP_005268678.1 12 1275
          XM_006719224.2 XP_006719287.1 8 1136
          XM_011537855.2 XP_011536157.1 9 794
          XM_011537856.2 XP_011536158.1 10 1212
          XM_011537857.2 XP_011536159.1 12 1266
          XM_017018768.1 XP_016874257.1 9 809
          XM_017018769.1 XP_016874258.1 10 1180
          GenBank mRNA AB040884.3 - 9 848
          AF323730.1 - 8 1131
          AF392452.1 - 9 1178
          AK055835.1 - 8 729
          AK092160.1 - 6 1063
          AK289997.1 - 9 897
          BC082235.1 - 9 1116
          BC093834.1 - 8 776 7939679
          BC101529.1 - 8 776 8069035
          BC111728.1 - 9 1071 40029118
          BX647211.1 - 8 733
          CR749542.1 - 11 981

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00368264_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of sequestering of triglyceride
          phospholipid transport
          negative regulation of cell migration
          activation of protein kinase B activity
          fat cell differentiation
          positive regulation of insulin receptor signaling pathway
          positive regulation of protein kinase B signaling
          protein localization to nuclear pore
          positive regulation of glucose import in response to insulin stimulus
          phosphatidylserine binding
          phospholipid transporter activity
          cholesterol binding
          phosphatidylinositol-4-phosphate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fxlmx:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0