Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030945.3 | NP_112207.1 | 5, 6 | 730 | ||
NM_181435.5 | NP_852100.3 | 5, 6 | 949 | |||
GenBank mRNA | AF173888.1 | - | 5, 6 | 632 | ||
AF326976.1 | - | 5, 6 | 665 | |||
AF329837.1 | - | 5, 6 | 672 | |||
AK075533.1 | - | 5, 6 | 677 | |||
AK127389.1 | - | 1, 2 | 518 | |||
AK295968.1 | - | 5, 6 | 949 | |||
AK315921.1 | - | 5, 6 | 936 | |||
AY358388.1 | - | 5, 6 | 665 | |||
BC016021.1 | - | 2, 3 | 366 | |||
BC112925.1 | - | 5, 6 | 604 | 5199982 | ||
BC120990.2 | - | 5, 6 | 879 | 40116366 | ||
BX640995.1 | - | 6, 7 | 552 | |||
EU399231.1 | - | 5, 6 | 594 | |||
EU399232.1 | - | 5, 6 | 813 |
Gene Symbol: C1QTNF3-AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037951.1 | 5, 6 | 616 | |||
GenBank mRNA | AB209261.1 | - | 5, 6 | 625 |