Search
Search

VASN| Gene Symbol: VASN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138440.2 | NP_612449.2 | 2 | 1639 | ||
| GenBank mRNA | AY166584.1 | - | 1 | 1484 | ||
| AY358298.1 | - | 3 | 1395 | |||
| AY358299.1 | - | 2 | 1603 | |||
| BC013767.1 | - | 1 | 1268 | |||
| BC068575.1 | - | 2 | 1639 | 30349094 | ||