Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135556.1 | NP_001129028.1 | 3 | 426 | ||
NM_001135557.1 | NP_001129029.1 | 3 | 426 | |||
NM_001278421.1 | NP_001265350.1 | 3 | 426 | |||
NM_001278422.1 | NP_001265351.1 | 3 | 426 | |||
NM_004411.4 | NP_004402.1 | 3 | 426 | |||
XM_011515861.1 | XP_011514163.1 | 3 | 273 | |||
XM_011515862.1 | XP_011514164.1 | 3 | 273 | |||
XM_011515863.2 | XP_011514165.1 | 3 | 380 | |||
XM_017011804.1 | XP_016867293.1 | 3 | 1013 | |||
XM_017011805.1 | XP_016867294.1 | 3 | 251 | |||
XM_017011806.1 | XP_016867295.1 | 3 | 426 | |||
XM_017011807.1 | XP_016867296.1 | 3 | 380 | |||
GenBank mRNA | AF063228.1 | - | 3 | 336 | ||
AF123074.1 | - | 3 | 335 | |||
AK091339.1 | - | 4 | 326 | |||
AK297427.1 | - | 3 | 338 | |||
AK309419.1 | - | 3 | 249 | |||
AK313148.1 | - | 3 | 373 | |||
BC022540.1 | - | 3 | 335 | 4793415 | ||
BC032945.2 | - | 3 | 378 | 4801387 | ||
BC037854.1 | - | 3 | 368 | 4817657 | ||
BC053542.1 | - | 3 | 366 | 5727968 | ||
DA491512.1 | - | 3 | 426 |