Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039589.1 | NP_001034678.1 | 3 | 502 | ||
NM_001319165.1 | NP_001306094.1 | 3 | 492 | |||
NM_001319166.1 | NP_001306095.1 | 3 | 502 | |||
NM_001319167.1 | NP_001306096.1 | 3 | 559 | |||
NM_001319168.1 | NP_001306097.1 | 3 | 450 | |||
NM_001319170.1 | NP_001306099.1 | 3 | 562 | |||
NM_001319171.1 | NP_001306100.1 | 3 | 502 | |||
NM_001384.4 | NP_001375.2 | 3 | 502 | |||
XM_017000502.1 | XP_016855991.1 | 3 | 504 | |||
XM_017000503.1 | XP_016855992.1 | 3 | 438 | |||
XM_017000504.1 | XP_016855993.1 | 3 | 546 | |||
XM_017000505.1 | XP_016855994.1 | 3 | 478 | |||
XM_017000506.1 | XP_016855995.1 | 3 | 546 | |||
GenBank mRNA | AK091303.1 | - | 3 | 421 | ||
AK315506.1 | - | 3 | 486 | |||
BC001389.2 | - | 3 | 461 | 3051455 | ||
BC003181.1 | - | 3 | 479 | 2958202 | ||
BC016956.1 | - | 3 | 461 | 3900854 | ||
BG385934.1 | - | 3 | 487 | |||
BG472282.1 | - | 3 | 520 | |||
BG747150.1 | - | 3 | 539 | |||
BM564151.1 | - | 453 | ||||
BM794530.1 | - | 3 | 444 | |||
BQ056213.1 | - | 3 | 476 | |||
BQ215136.1 | - | 3 | 510 | |||
BT007431.1 | - | 3 | 307 | |||
CX164803.1 | - | 3 | 429 | |||
DA906783.1 | - | 3 | 504 |