Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145203.5 | NP_660204.2 | 1 | 601 | ||
GenBank mRNA | BC028723.2 | - | 1 | 603 | 5266487 |