Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DRD1 SIRNA Assays ›
          Gene Symbol:
          DRD1
          Gene Name:
          dopamine receptor D1
          Gene Aliases:
          DADR, DRD1A
          Chromosome Location:
          Chr.5: 175440672 - 175444160 on Build GRCh38
          UniGene ID:
          Hs.2624
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4283

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DRD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000794.3 NP_000785.1 2 1707
          GenBank mRNA AF498961.1 - 1 762
          AK314031.1 - 1 1193
          BC074978.2 - 1 1038 30915514
          BC074979.2 - 1 1038 30915297
          BC096837.1 - 1 765 6519220
          CR541922.1 - 1 762
          X58987.1 - 1 1038

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          conditioned taste aversion
          behavioral fear response
          synaptic transmission, dopaminergic
          response to amphetamine
          negative regulation of protein kinase activity
          protein import into nucleus
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          adenylate cyclase-activating dopamine receptor signaling pathway
          synapse assembly
          learning
          memory
          mating behavior
          grooming behavior
          adult walking behavior
          visual learning
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          astrocyte development
          response to activity
          dopamine transport
          transmission of nerve impulse
          neuronal action potential
          regulation of vasoconstriction
          calcium-mediated signaling
          dentate gyrus development
          striatum development
          orbitofrontal cortex development
          cerebral cortex GABAergic interneuron migration
          positive regulation of cell migration
          negative regulation of cell migration
          peristalsis
          positive regulation of cAMP biosynthetic process
          response to food
          response to estradiol
          response to retinoic acid
          cellular response to insulin stimulus
          response to nicotine
          operant conditioning
          social behavior
          response to antidepressant
          regulation of dopamine metabolic process
          vasodilation
          negative regulation of circadian sleep/wake cycle, sleep
          dopamine metabolic process
          response to drug
          maternal behavior
          response to amino acid
          positive regulation of potassium ion transport
          response to morphine
          response to ethanol
          positive regulation of membrane potential
          glucose import
          habituation
          sensitization
          behavioral response to cocaine
          regulation of long-term neuronal synaptic plasticity
          response to steroid hormone
          positive regulation of release of sequestered calcium ion into cytosol
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          regulation of dopamine uptake involved in synaptic transmission
          positive regulation of synaptic transmission, glutamatergic
          prepulse inhibition
          phospholipase C-activating dopamine receptor signaling pathway
          long-term synaptic potentiation
          long term synaptic depression
          cellular response to hypoxia
          cellular response to catecholamine stimulus
          positive regulation of long-term synaptic potentiation
          positive regulation of feeding behavior
          dopamine neurotransmitter receptor activity, coupled via Gs
          G-protein alpha-subunit binding
          dopamine neurotransmitter receptor activity
          protein binding
          drug binding
          protein phosphatase binding
          angiotensin receptor binding
          D3 dopamine receptor binding
          protein complex binding
          dopamine binding
          protein heterodimerization activity
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0