Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122890.2 | NP_001116362.1 | 4 | 542 | ||
NM_001288702.1 | NP_001275631.1 | 4 | 560 | |||
NM_153338.3 | NP_699169.2 | 3 | 446 | |||
GenBank mRNA | AK074646.1 | - | 3 | 431 | ||
AK296645.1 | - | 4 | 541 | |||
AK298329.1 | - | 3 | 523 | |||
AK300648.1 | - | 4 | 520 | |||
BC063111.1 | - | 4 | 512 | 4593403 | ||
DA645854.1 | - | 3 | 431 | |||
DC345075.1 | - | 3 | 446 | |||
DC345192.1 | - | 4 | 539 |