Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145257.4 | NP_660300.3 | 3 | 600 | ||
GenBank mRNA | AK123465.1 | - | 2 | 389 | ||
AK291437.1 | - | 2 | 561 | |||
BC015419.1 | - | 2 | 574 | 4396825 | ||
BC060777.1 | - | 3 | 1505 | |||
BC071609.1 | - | 3 | 646 | 5264741 |