Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099783.1 | NP_001093253.1 | 5 | 398 | ||
NM_173487.2 | NP_775758.2 | 5 | 670 | |||
XM_005262734.4 | XP_005262791.1 | 5 | 665 | |||
XM_017007722.1 | XP_016863211.1 | 5 | 1727 | |||
XM_017007723.1 | XP_016863212.1 | 5 | 1477 | |||
XM_017007724.1 | XP_016863213.1 | 5 | 653 | |||
GenBank mRNA | AK091022.1 | - | 5 | 659 | ||
AK309229.1 | - | 5 | 644 | |||
BC016358.2 | - | 11 | ||||
BC032582.1 | - | 5 | 601 | 5494700 | ||
BE566349.1 | - | 5 | 353 | |||
BQ549846.1 | - | 5 | 429 | |||
BX538164.1 | - | 5 | 611 |