Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102609.2 | NP_001096079.1 | 4 | 301 | ||
NM_001305393.1 | NP_001292322.1 | 4 | 320 | |||
NM_001305394.1 | NP_001292323.1 | 4 | 309 | |||
NM_001305395.1 | NP_001292324.1 | 5 | 475 | |||
NM_001305396.1 | NP_001292325.1 | 5 | 464 | |||
NM_001305397.1 | NP_001292326.1 | 5 | 448 | |||
XM_011534432.2 | XP_011532734.1 | 4 | 772 | |||
XM_017009029.1 | XP_016864518.1 | 5 | 1662 | |||
XM_017009030.1 | XP_016864519.1 | 5 | 1634 | |||
XM_017009031.1 | XP_016864520.1 | 4 | 361 | |||
XM_017009032.1 | XP_016864521.1 | 4 | 285 | |||
XM_017009033.1 | XP_016864522.1 | 4 | 352 | |||
GenBank mRNA | BC092511.1 | - | 3 | 1281 | ||
BU561730.1 | - | 5 | 411 | |||
BX100176.1 | - | 3 | 221 | |||
HY000873.1 | - | 4 | 387 | |||
HY001392.1 | - | 4 | 371 | |||
HY005987.1 | - | 4 | 320 | |||
HY016595.1 | - | 4 | 226 | |||
HY038777.1 | - | 4 | 301 |