Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178829.4 | NP_849151.2 | 2 | 741 | ||
GenBank mRNA | AB083484.1 | - | 2 | 689 | ||
BC014596.1 | - | 2 | 746 | 4825428 |