Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTCFL SIRNA Assays ›
Gene Symbol:
CTCFL
Gene Name:
CCCTC-binding factor like
Gene Aliases:
BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2
Chromosome Location:
Chr.20: 57495965 - 57525652 on Build GRCh38
UniGene ID:
Hs.131543
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s44349
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTCFL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001269040.1
NP_001255969.1
9
1799
NM_001269041.1
NP_001255970.1
8
2271
NM_001269042.1
NP_001255971.1
9
1799
NM_001269043.1
NP_001255972.1
9
1720
NM_001269044.1
NP_001255973.1
8
2271
NM_001269045.1
NP_001255974.1
9
1799
NM_001269046.1
NP_001255975.1
8
2095
NM_001269047.1
NP_001255976.1
9
1799
NM_001269048.1
NP_001255977.1
5
1839
NM_001269049.1
NP_001255978.1
8
1166
NM_001269050.1
NP_001255979.1
8
1146
NM_001269054.1
NP_001255983.1
7
955
NM_001269055.1
NP_001255984.1
7
935
NM_080618.3
NP_542185.2
10
1899
NR_072975.1
9
2908
GenBank mRNA
AF336042.1
-
9
1698
AK128070.1
-
9
1802
AK313990.1
-
9
1733
AY071919.1
-
9
1726
BC130486.1
-
9
1676
40146517
DQ294738.1
-
8
2020
DQ778108.1
-
9
1799
DQ778109.1
-
10
1899
DQ778110.1
-
8
2271
DQ778111.1
-
9
1698
DQ778112.1
-
8
2095
DQ778115.2
-
8
2271
DQ778116.2
-
6
1876
DQ778118.2
-
9
2908
DQ778121.3
-
5
1839
DQ778122.2
-
9
1799
DQ778123.2
-
9
1799
DQ778124.2
-
7
934
DQ778125.2
-
8
1145
DQ778126.2
-
8
1144
DQ778127.2
-
7
933
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression by genetic imprinting
transcription from RNA polymerase II promoter
cell cycle
positive regulation of gene expression
histone methylation
DNA methylation involved in gamete generation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of histone H3-K4 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
histone binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0