Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIB2 SIRNA Assays ›
Gene Symbol:
MIB2
Gene Name:
mindbomb E3 ubiquitin protein ligase 2
Gene Aliases:
ZZANK1, ZZZ5
Chromosome Location:
Chr.1: 1615188 - 1630610 on Build GRCh38
UniGene ID:
Hs.135805
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s44470
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170686.1
NP_001164157.1
19
2986
NM_001170687.1
NP_001164158.1
19
2956
NM_001170688.1
NP_001164159.1
18
2803
NM_001170689.1
NP_001164160.1
17
2596
NM_080875.2
NP_543151.2
19
2998
NR_033183.1
19
2781
XM_006710372.1
XP_006710435.1
18
2862
XM_011540731.2
XP_011539033.1
20
3834
XM_011540736.2
XP_011539038.1
20
3735
XM_011540737.2
XP_011539039.1
19
3672
XM_011540741.2
XP_011539043.1
18
3567
XM_011540742.2
XP_011539044.1
18
3564
XM_017000349.1
XP_016855838.1
19
3906
XM_017000350.1
XP_016855839.1
19
3894
XM_017000351.1
XP_016855840.1
19
3888
XM_017000352.1
XP_016855841.1
19
3864
XM_017000353.1
XP_016855842.1
18
3843
XM_017000354.1
XP_016855843.1
19
3807
XM_017000355.1
XP_016855844.1
17
3738
XM_017000356.1
XP_016855845.1
17
3735
XM_017000357.1
XP_016855846.1
18
3711
XM_017000358.1
XP_016855847.1
19
3204
XM_017000359.1
XP_016855848.1
19
3192
XM_017000360.1
XP_016855849.1
19
3151
XM_017000361.1
XP_016855850.1
18
3042
XM_017000362.1
XP_016855851.1
19
3244
XM_017000363.1
XP_016855852.1
19
3207
XM_017000364.1
XP_016855853.1
17
2847
XM_017000365.1
XP_016855854.1
19
2934
XM_017000366.1
XP_016855855.1
18
2871
GenBank mRNA
AB074480.1
-
19
2909
AB076691.1
-
19
2867
AB076692.1
-
18
2714
AB076693.1
-
19
2897
AB097000.1
-
19
2832
AK000655.1
-
2
251
AK091610.1
-
13
1802
AK095914.1
-
13
1804
AK096295.1
-
14
1909
AK097106.1
-
15
2228
AK098785.1
-
20
2784
AK122751.1
-
19
2700
AK122863.1
-
18
2992
AK128167.1
-
17
2556
AK128553.1
-
18
3912
AK293642.1
-
18
2978
AK302757.1
-
19
2950
AL834527.1
-
14
2030
BC016490.1
-
19
2835
BC037542.1
-
19
2878
5168033
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0