Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPAR1 SIRNA Assays ›
Gene Symbol:
LPAR1
Gene Name:
lysophosphatidic acid receptor 1
Gene Aliases:
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location:
Chr.9: 110873252 - 111039259 on Build GRCh38
UniGene ID:
Hs.126667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4450
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001401.3
NP_001392.2
3
1111
NM_057159.2
NP_476500.1
4
1189
XM_005251781.3
XP_005251838.1
4
1334
XM_005251782.3
XP_005251839.1
4
1428
XM_017014383.1
XP_016869872.1
4
1228
XM_017014384.1
XP_016869873.1
5
1602
XM_017014385.1
XP_016869874.1
6
1195
XM_017014386.1
XP_016869875.1
6
1495
XM_017014387.1
XP_016869876.1
6
1231
XM_017014388.1
XP_016869877.1
6
1414
XM_017014389.1
XP_016869878.1
5
1157
XM_017014390.1
XP_016869879.1
6
1192
XM_017014391.1
XP_016869880.1
6
1216
XM_017014392.1
XP_016869881.1
5
1459
XM_017014393.1
XP_016869882.1
6
1255
XM_017014394.1
XP_016869883.1
5
1384
XM_017014395.1
XP_016869884.1
5
1212
XM_017014396.1
XP_016869885.1
4
1154
XM_017014397.1
XP_016869886.1
5
1085
XM_017014398.1
XP_016869887.1
5
1328
XM_017014399.1
XP_016869888.1
5
1083
XM_017014400.1
XP_016869889.1
6
1444
XM_017014401.1
XP_016869890.1
5
1115
XM_017014402.1
XP_016869891.1
5
1172
XM_017014403.1
XP_016869892.1
6
1170
XM_017014404.1
XP_016869893.1
4
1368
XM_017014405.1
XP_016869894.1
6
1357
XM_017014406.1
XP_016869895.1
5
1091
XM_017014407.1
XP_016869896.1
5
1147
GenBank mRNA
AK022808.1
-
3
1111
AK290287.1
-
5
1464
AK296374.1
-
2
792
AK298590.1
-
2
768
AK299279.1
-
2
842
AY322546.1
-
2
763
BC030615.2
-
4
1018
4815446
BC036034.1
-
4
1148
5272431
BC073167.1
-
5
1058
6193969
U78192.1
-
3
885
U80811.1
-
2
789
Y09479.1
-
3
885
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
regulation of cell shape
negative regulation of neuron projection development
oligodendrocyte development
cerebellum development
optic nerve development
corpus callosum development
negative regulation of cAMP biosynthetic process
bleb assembly
positive regulation of Rho protein signal transduction
myelination
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of stress fiber assembly
cell chemotaxis
positive regulation of dendritic spine development
cellular response to oxygen levels
positive regulation of smooth muscle cell chemotaxis
cellular response to 1-oleoyl-sn-glycerol 3-phosphate
G-protein alpha-subunit binding
G-protein coupled receptor activity
protein binding
PDZ domain binding
lysophosphatidic acid binding
lysophosphatidic acid receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-njplc:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0