Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FRMPD2 SIRNA Assays ›
          Gene Symbol:
          FRMPD2 FRMPD2B
          Gene Name:
          FERM and PDZ domain containing 2
          FERM and PDZ domain containing 2B, pseudogene
          Gene Aliases:
          PDZD5C, PDZK4, PDZK5C
          FRMPD2L1, FRMPD2L2, FRMPD2P1, FRMPD2P2, FRMPDP2, PDZD5A, PDZD5B, PDZK5A, PDZK5B, bA556L1.2, yX59F3.2
          Chromosome Location:
          Chr.10: 48156559 - 48274898 on Build GRCh38
          Chromosome Location:
          Chr.10: 46876674 - 46895326 on Build GRCh38
          UniGene ID:
          Hs.664786
          Hs.664786
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s44508

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FRMPD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018071.3 NP_001018081.3 27, 28 3873
          NM_001042512.2 NP_001035977.2 4, 5 741
          NM_001318191.1 NP_001305120.1 25, 26 3514
          XM_017015744.1 XP_016871233.1 4, 5 557
          GenBank mRNA AK123038.1 - 25, 26 3514
          AY491519.1 - 4, 5 740
          BC031614.1 - 6, 7 736
          BC073954.1 - 4, 5 722
          BC144636.1 - 4, 5 682 9053167
          BC144639.1 - 4, 5 682 9053170
          CR749241.1 - 27, 28 3845
          Gene Symbol: FRMPD2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033172.1 4, 5 795
          GenBank mRNA AK294650.1 - 4, 5 750
          AK313659.1 - 4, 5 752
          AK315971.1 - 4, 5 795
          BC071635.1 - 4, 5 707
          BC072395.1 - 4, 5 721
          BC110798.1 - 4, 5 718
          BC137431.1 - 4, 5 682 9021429
          BC137434.1 - 4, 5 682 9021432
          BC137439.1 - 4, 5 680 9021437
          BC137440.1 - 4, 5 682 9021438
          BC144635.1 - 4, 5 682 9053166

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          bicellular tight junction assembly
          protein binding
          1-phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0