Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPAR1 SIRNA Assays ›
Gene Symbol:
LPAR1
Gene Name:
lysophosphatidic acid receptor 1
Gene Aliases:
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location:
Chr.9: 110873252 - 111039259 on Build GRCh38
UniGene ID:
Hs.126667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4451
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPAR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001401.3
NP_001392.2
3
919
NM_057159.2
NP_476500.1
4
997
XM_005251781.3
XP_005251838.1
4
1142
XM_005251782.3
XP_005251839.1
4
1236
XM_017014383.1
XP_016869872.1
4
1036
XM_017014384.1
XP_016869873.1
5
1410
XM_017014385.1
XP_016869874.1
6
1003
XM_017014386.1
XP_016869875.1
6
1303
XM_017014387.1
XP_016869876.1
6
1039
XM_017014388.1
XP_016869877.1
6
1222
XM_017014389.1
XP_016869878.1
5
965
XM_017014390.1
XP_016869879.1
6
1000
XM_017014391.1
XP_016869880.1
6
1024
XM_017014392.1
XP_016869881.1
5
1267
XM_017014393.1
XP_016869882.1
6
1063
XM_017014394.1
XP_016869883.1
5
1192
XM_017014395.1
XP_016869884.1
5
1020
XM_017014396.1
XP_016869885.1
4
962
XM_017014397.1
XP_016869886.1
5
893
XM_017014398.1
XP_016869887.1
5
1136
XM_017014399.1
XP_016869888.1
5
891
XM_017014400.1
XP_016869889.1
6
1252
XM_017014401.1
XP_016869890.1
5
923
XM_017014402.1
XP_016869891.1
5
980
XM_017014403.1
XP_016869892.1
6
978
XM_017014404.1
XP_016869893.1
4
1176
XM_017014405.1
XP_016869894.1
6
1165
XM_017014406.1
XP_016869895.1
5
899
XM_017014407.1
XP_016869896.1
5
955
GenBank mRNA
AK022808.1
-
3
919
AK290287.1
-
5
1272
AK296374.1
-
2
600
AK298590.1
-
2
576
AK299279.1
-
2
650
AY322546.1
-
2
571
BC030615.2
-
4
826
4815446
BC036034.1
-
4
956
5272431
BC073167.1
-
5
866
6193969
U78192.1
-
3
693
U80811.1
-
2
597
Y09479.1
-
3
693
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
activation of phospholipase C activity
positive regulation of cytosolic calcium ion concentration
regulation of cell shape
negative regulation of neuron projection development
oligodendrocyte development
cerebellum development
optic nerve development
corpus callosum development
negative regulation of cAMP biosynthetic process
bleb assembly
positive regulation of Rho protein signal transduction
myelination
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of stress fiber assembly
cell chemotaxis
positive regulation of dendritic spine development
cellular response to oxygen levels
positive regulation of smooth muscle cell chemotaxis
cellular response to 1-oleoyl-sn-glycerol 3-phosphate
G-protein alpha-subunit binding
G-protein coupled receptor activity
protein binding
PDZ domain binding
lysophosphatidic acid binding
lysophosphatidic acid receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0