Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LPAR1 SIRNA Assays ›
          Gene Symbol:
          LPAR1
          Gene Name:
          lysophosphatidic acid receptor 1
          Gene Aliases:
          EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
          Chromosome Location:
          Chr.9: 110873252 - 111039259 on Build GRCh38
          UniGene ID:
          Hs.126667
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4452

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LPAR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001401.3 NP_001392.2 3 698
          NM_057159.2 NP_476500.1 4 776
          XM_005251781.3 XP_005251838.1 4 921
          XM_005251782.3 XP_005251839.1 4 1015
          XM_017014383.1 XP_016869872.1 4 815
          XM_017014384.1 XP_016869873.1 5 1189
          XM_017014385.1 XP_016869874.1 6 782
          XM_017014386.1 XP_016869875.1 6 1082
          XM_017014387.1 XP_016869876.1 6 818
          XM_017014388.1 XP_016869877.1 6 1001
          XM_017014389.1 XP_016869878.1 5 744
          XM_017014390.1 XP_016869879.1 6 779
          XM_017014391.1 XP_016869880.1 6 803
          XM_017014392.1 XP_016869881.1 5 1046
          XM_017014393.1 XP_016869882.1 6 842
          XM_017014394.1 XP_016869883.1 5 971
          XM_017014395.1 XP_016869884.1 5 799
          XM_017014396.1 XP_016869885.1 4 741
          XM_017014397.1 XP_016869886.1 5 672
          XM_017014398.1 XP_016869887.1 5 915
          XM_017014399.1 XP_016869888.1 5 670
          XM_017014400.1 XP_016869889.1 6 1031
          XM_017014401.1 XP_016869890.1 5 702
          XM_017014402.1 XP_016869891.1 5 759
          XM_017014403.1 XP_016869892.1 6 757
          XM_017014404.1 XP_016869893.1 4 955
          XM_017014405.1 XP_016869894.1 6 944
          XM_017014406.1 XP_016869895.1 5 678
          XM_017014407.1 XP_016869896.1 5 734
          GenBank mRNA AK022808.1 - 3 698
          AK290287.1 - 5 1051
          AK296374.1 - 2 379
          AK298590.1 - 2 355
          AK299279.1 - 2 429
          AY322546.1 - 2 350
          BC030615.2 - 4 605 4815446
          BC036034.1 - 4 735 5272431
          BC073167.1 - 5 645 6193969
          U80811.1 - 2 376

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          activation of phospholipase C activity
          positive regulation of cytosolic calcium ion concentration
          regulation of cell shape
          negative regulation of neuron projection development
          oligodendrocyte development
          cerebellum development
          optic nerve development
          corpus callosum development
          negative regulation of cAMP biosynthetic process
          bleb assembly
          positive regulation of Rho protein signal transduction
          myelination
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of MAPK cascade
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          positive regulation of stress fiber assembly
          cell chemotaxis
          positive regulation of dendritic spine development
          cellular response to oxygen levels
          positive regulation of smooth muscle cell chemotaxis
          cellular response to 1-oleoyl-sn-glycerol 3-phosphate
          G-protein alpha-subunit binding
          G-protein coupled receptor activity
          protein binding
          PDZ domain binding
          lysophosphatidic acid binding
          lysophosphatidic acid receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-r46r8:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0