Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EDN1 SIRNA Assays ›
          Gene Symbol:
          EDN1
          Gene Name:
          endothelin 1
          Gene Aliases:
          ARCND3, ET1, HDLCQ7, PPET1, QME
          Chromosome Location:
          Chr.6: 12256464 - 12297194 on Build GRCh38
          UniGene ID:
          Hs.511899
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4457

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EDN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001168319.1 NP_001161791.1 4 844
          NM_001955.4 NP_001946.3 4 847
          XM_011514330.2 XP_011512632.1 5 603
          XM_011514331.2 XP_011512633.1 6 764
          XM_011514332.2 XP_011512634.1 5 601
          XM_017010331.1 XP_016865820.1 5 787
          GenBank mRNA AK226096.1 - 4 762
          AK291838.1 - 4 784
          AK313914.1 - 4 779
          BC009720.1 - 4 716 3900704
          BX439515.2 - 4 807
          CR541903.1 - 4 513
          S56805.1 - 4 849
          Y00749.1 - 4 765

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          prostaglandin biosynthetic process
          patterning of blood vessels
          in utero embryonic development
          histamine secretion
          regulation of systemic arterial blood pressure by endothelin
          regulation of pH
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          cell-cell signaling
          heart development
          respiratory gaseous exchange
          body fluid secretion
          positive regulation of cell proliferation
          dorsal/ventral pattern formation
          response to ozone
          multicellular organism aging
          positive regulation of heart rate
          positive regulation of endothelial cell migration
          positive regulation of cardiac muscle hypertrophy
          positive regulation of receptor biosynthetic process
          neural crest cell development
          phosphatidylinositol 3-kinase signaling
          response to activity
          artery smooth muscle contraction
          vein smooth muscle contraction
          glucose transport
          cell growth
          regulation of vasoconstriction
          sensory perception of pain
          calcium-mediated signaling
          peptide hormone secretion
          nitric oxide transport
          negative regulation of blood coagulation
          positive regulation of cell migration
          neutrophil chemotaxis
          negative regulation of cAMP biosynthetic process
          phospholipase D-activating G-protein coupled receptor signaling pathway
          negative regulation of cellular protein metabolic process
          positive regulation of prostaglandin secretion
          response to lipopolysaccharide
          response to testosterone
          negative regulation of smooth muscle cell apoptotic process
          response to prostaglandin F
          response to nicotine
          cellular response to drug
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          response to muscle stretch
          epithelial fluid transport
          vasoconstriction
          protein kinase C deactivation
          middle ear morphogenesis
          positive regulation of odontogenesis
          superoxide anion generation
          rhythmic excitation
          response to amino acid
          positive regulation of MAP kinase activity
          positive regulation of JUN kinase activity
          response to leptin
          leukocyte activation
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell size
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle contraction
          positive regulation of hormone secretion
          negative regulation of hormone secretion
          inositol phosphate-mediated signaling
          positive regulation of smooth muscle cell proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          cartilage development
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          negative regulation of nitric-oxide synthase biosynthetic process
          membrane depolarization
          regulation of sensory perception of pain
          maternal process involved in parturition
          positive regulation of sarcomere organization
          positive regulation of prostaglandin-endoperoxide synthase activity
          positive regulation of cell growth involved in cardiac muscle cell development
          positive regulation of chemokine-mediated signaling pathway
          cellular response to calcium ion
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to peptide hormone stimulus
          cellular response to glucocorticoid stimulus
          cellular response to mineralocorticoid stimulus
          cellular response to fatty acid
          cellular response to hypoxia
          response to dexamethasone
          cellular response to transforming growth factor beta stimulus
          response to salt
          cytokine activity
          hormone activity
          protein binding
          endothelin A receptor binding
          endothelin B receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0